Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IP6K3
Entrez Gene ID
101118907
Gene Name
inositol hexakisphosphate kinase 3
Gene Aliases
-
Chromosome Location
Chr.20: 9546154 - 9570320 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959002.1 XP_027814803.1 4 - 5 951 55

View Full Product Details