Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRSF3
Entrez Gene ID
100135442
Gene Name
serine and arginine rich splicing factor 3
Gene Aliases
SFRS3
Chromosome Location
Chr.20: 12162424 - 12170011 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195316.1 NP_001182245.1 1 - 2 207 93
XM_012100410.2 XP_011955800.1 2 - 3 340 93
GenBank mRNA EU370536.1 - 1 - 2 207 93

View Full Product Details