Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SS18L1
Entrez Gene ID
101113515
Gene Name
SS18L1 subunit of BAF chromatin remodeling complex
Gene Aliases
-
Chromosome Location
Chr.13: 57376707 - 57404471 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976979.1 XP_027832780.1 5 - 6 632 73

View Full Product Details