Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATAD1
Entrez Gene ID
105609643
Gene Name
GATA zinc finger domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 11408529 - 11415709 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968441.1 XP_027824242.1 2 - 3 433 86

View Full Product Details