Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101104950
Entrez Gene ID
101104950
Gene Name
synaptotagmin-4
Gene Aliases
SYT4
Chromosome Location
Chr.23: 14387340 - 14397438 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020409.4 XP_004020458.1 3 - 4 1216 66

View Full Product Details