Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUP58
Entrez Gene ID
101115885
Gene Name
nucleoporin 58
Gene Aliases
NUPL1
Chromosome Location
Chr.10: 35874918 - 35904929 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027973654.1 XP_027829455.1 13 - 14 1681 64
XM_027973656.1 XP_027829457.1 13 - 14 1576 64

View Full Product Details