Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT1
Entrez Gene ID
101103389
Gene Name
Wnt family member 1
Gene Aliases
-
Chromosome Location
Chr.3: 147154121 - 147157129 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006386.4 XP_004006435.2 3 - 4 623 85

View Full Product Details