Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD8
Entrez Gene ID
101119822
Gene Name
potassium channel tetramerization domain containing 8
Gene Aliases
-
Chromosome Location
Chr.6: 68961914 - 69243985 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004009799.4 XP_004009848.1 1 - 2 992 72

View Full Product Details