Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HYPK
Entrez Gene ID
101111231
Gene Name
huntingtin interacting protein K
Gene Aliases
-
Chromosome Location
Chr.18: 55744891 - 55747167 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957323.1 XP_027813124.1 2 - 3 1260 62

View Full Product Details