Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP2
Entrez Gene ID
101117688
Gene Name
bone morphogenetic protein 2
Gene Aliases
-
Chromosome Location
Chr.13: 51168039 - 51180602 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004014353.4 XP_004014402.1 2 - 3 1512 56

View Full Product Details