Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL12A
Entrez Gene ID
443064
Gene Name
interleukin 12A
Gene Aliases
IL12
Chromosome Location
Chr.1: 248087948 - 248094879 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009736.1 NP_001009736.1 2 - 3 169 111
XM_012146332.3 XP_012001722.2 1 - 2 279 111
GenBank mRNA AF173557.1 - 2 - 3 169 111

View Full Product Details