Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OOEP
Entrez Gene ID
101107104
Gene Name
oocyte expressed protein
Gene Aliases
-
Chromosome Location
Chr.9: 314850 - 315978 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287467.1 NP_001274396.1 1 - 2 203 93
GenBank mRNA KF218578.1 - 1 - 2 203 93

View Full Product Details