Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
C4BPB
Entrez Gene ID
101114204
Gene Name
complement component 4 binding protein beta
Gene Aliases
-
Chromosome Location
Chr.12: 4532631 - 4545325 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308575.1 NP_001295504.1 3 - 4 327 127
XM_027975563.1 XP_027831364.1 4 - 5 632 127
XM_027975564.1 XP_027831365.1 3 - 4 547 127
GenBank mRNA HF674450.1 - 3 - 4 327 127

View Full Product Details