Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDR16C5
Entrez Gene ID
100302354
Gene Name
short chain dehydrogenase/reductase family 16C member 5
Gene Aliases
-
Chromosome Location
Chr.9: 39168621 - 39184960 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163052.1 NP_001156524.1 2 - 3 470 105
XM_012183366.3 XP_012038756.1 3 - 4 578 105
GenBank mRNA FJ958348.1 - 2 - 3 470 105

View Full Product Details