Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC100527962
Entrez Gene ID
100527962
Gene Name
KRAB-domain containing protein
Gene Aliases
-
Chromosome Location
Chr.25: 12409610 - 12451644 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199670.1 NP_001186599.1 3 - 4 369 90
GenBank mRNA HM991147.1 - 3 - 4 369 90

View Full Product Details