Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGN
Entrez Gene ID
100171395
Gene Name
regucalcin
Gene Aliases
-
Chromosome Location
Chr.X: 59998047 - 60013640 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130935.1 NP_001124407.1 3 - 4 567 77
XM_012106097.2 XP_011961487.1 4 - 5 607 77
XM_012106098.3 XP_011961488.1 4 - 5 666 77
XM_012106099.3 XP_011961489.1 4 - 5 671 77
XM_012106101.3 XP_011961491.1 4 - 5 610 77
XM_012106102.3 XP_011961492.1 4 - 5 684 77
XM_027962744.1 XP_027818545.1 4 - 5 671 77
GenBank mRNA EU797609.1 - 3 - 4 567 77

View Full Product Details