Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RETN
Entrez Gene ID
101122857
Gene Name
resistin
Gene Aliases
-
Chromosome Location
Chr.5: 14825961 - 14827474 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306111.1 NP_001293040.1 1 - 2 115 72
XM_027969185.1 XP_027824986.1 2 - 3 366 72
GenBank mRNA KJ704841.1 - 1 - 2 115 72

View Full Product Details