Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101109717
Entrez Gene ID
101109717
Gene Name
protein Mis18-alpha-like
Gene Aliases
-
Chromosome Location
Chr.10: 29561324 - 29614727 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027973839.1 XP_027829640.1 3 - 3 714 73

Gene Symbol
LOC114112857
Entrez Gene ID
114112857
Gene Name
protein Mis18-alpha
Gene Aliases
-
Chromosome Location
Chr.2: 136580880 - 136581678 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963953.1 XP_027819754.1 1 - 1 399 73

Gene Symbol
MIS18A
Entrez Gene ID
101113820
Gene Name
MIS18 kinetochore protein A
Gene Aliases
-
Chromosome Location
Chr.1: 131425426 - 131438745 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978380.1 XP_027834181.1 2 - 3 407 73

View Full Product Details