Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
HES7
Entrez Gene ID
101121950
Gene Name
hes family bHLH transcription factor 7
Gene Aliases
-
Chromosome Location
Chr.11: 35659893 - 35662648 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974932.1 XP_027830733.1 1 - 2 323 88
XM_027974933.1 XP_027830734.1 1 - 2 323 88

View Full Product Details