Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRIP2
Entrez Gene ID
101118863
Gene Name
nuclear receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 227738648 - 227746632 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006958.4 XP_004007007.4 4 - 5 733 75

View Full Product Details