Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR3G
Entrez Gene ID
101106586
Gene Name
RNA polymerase III subunit G
Gene Aliases
-
Chromosome Location
Chr.5: 95009860 - 95063950 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096190.2 XP_014951676.2 3 - 4 2835 111
XM_027970507.1 XP_027826308.1 3 - 4 393 111
XM_027970508.1 XP_027826309.1 3 - 4 393 111
XM_027970509.1 XP_027826310.1 3 - 4 393 111
XM_027970510.1 XP_027826311.1 3 - 4 393 111

View Full Product Details