Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VTCN1
Entrez Gene ID
101107915
Gene Name
V-set domain containing T cell activation inhibitor 1
Gene Aliases
-
Chromosome Location
Chr.1: 99401548 - 99488663 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012179414.3 XP_012034804.2 2 - 3 165 79

View Full Product Details