Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AQP11
Entrez Gene ID
101111499
Gene Name
aquaporin 11
Gene Aliases
-
Chromosome Location
Chr.21: 20061777 - 20074388 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019435.4 XP_004019484.1 2 - 3 738 137

View Full Product Details