Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJB5
Entrez Gene ID
101119367
Gene Name
DnaJ heat shock protein family (Hsp40) member B5
Gene Aliases
-
Chromosome Location
Chr.2: 56174336 - 56183102 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964461.1 XP_027820262.1 1 - 2 189 102
XM_027964462.1 XP_027820263.1 2 - 3 397 102

View Full Product Details