Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP1
Entrez Gene ID
101119790
Gene Name
THAP domain containing 1
Gene Aliases
-
Chromosome Location
Chr.26: 40910163 - 40920858 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962556.1 XP_027818357.1 2 - 3 2045 117

View Full Product Details