Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LSM12
Entrez Gene ID
101113160
Gene Name
LSM12 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 19156109 - 19176201 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012976.4 XP_004013025.1 3 - 4 437 93

View Full Product Details