Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMD10
Entrez Gene ID
101116077
Gene Name
sterile alpha motif domain containing 10
Gene Aliases
-
Chromosome Location
Chr.13: 56171505 - 56176053 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976932.1 XP_027832733.1 2 - 3 326 53

View Full Product Details