Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAX1
Entrez Gene ID
101121136
Gene Name
ventral anterior homeobox 1
Gene Aliases
-
Chromosome Location
Chr.22: 40632668 - 40636755 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960403.1 XP_027816204.1 1 - 2 241 91

View Full Product Details