Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMC1
Entrez Gene ID
105603337
Gene Name
C-X9-C motif containing 1
Gene Aliases
-
Chromosome Location
Chr.19: 2887888 - 2967936 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015102208.2 XP_014957694.1 2 - 3 119 67
XM_015102210.2 XP_014957696.1 2 - 3 157 67

View Full Product Details