Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFX
Entrez Gene ID
101114932
Gene Name
zinc finger protein X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 23772991 - 23810535 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012106414.3 XP_011961804.1 6 - 7 1417 92
XM_012106416.3 XP_011961806.1 6 - 7 1157 92
XM_015104833.2 XP_014960319.1 6 - 7 1386 92
XM_027963307.1 XP_027819108.1 6 - 7 1780 92
XM_027963308.1 XP_027819109.1 6 - 7 1318 92

View Full Product Details