Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HECA
Entrez Gene ID
101122796
Gene Name
hdc homolog, cell cycle regulator
Gene Aliases
-
Chromosome Location
Chr.8: 70385654 - 70428878 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972500.1 XP_027828301.1 3 - 4 1599 74

View Full Product Details