Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF25
Entrez Gene ID
101116060
Gene Name
kinesin family member 25
Gene Aliases
-
Chromosome Location
Chr.8: 97102103 - 97122905 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972609.1 XP_027828410.1 8 - 9 1226 65

View Full Product Details