Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK12
Entrez Gene ID
101113003
Gene Name
kallikrein related peptidase 12
Gene Aliases
-
Chromosome Location
Chr.14: 60505219 - 60509859 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015398.4 XP_004015447.2 4 - 5 705 102

View Full Product Details