Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D1A
Entrez Gene ID
101117307
Gene Name
SH2 domain containing 1A
Gene Aliases
-
Chromosome Location
Chr.X: 116030731 - 116051386 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022366.4 XP_004022415.1 2 - 3 344 100

View Full Product Details