Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NDUFC1
- Entrez Gene ID
-
101116510
- Gene Name
- NADH:ubiquinone oxidoreductase subunit C1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 19898340 - 19902965 on Build Oar_rambouillet_v1.0
- Gene Symbol
- LOC114112897
- Entrez Gene ID
-
114112897
- Gene Name
- NADH dehydrogenase [ubiquinone] 1 subunit C1, mitochondrial
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 239912339 - 239912744 on Build Oar_rambouillet_v1.0
View Full Product Details