Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JTB
Entrez Gene ID
101102365
Gene Name
jumping translocation breakpoint
Gene Aliases
-
Chromosome Location
Chr.1: 110512133 - 110514265 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975544.1 XP_027831345.1 4 - 5 632 91

View Full Product Details