Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
ZIC4
Entrez Gene ID
101102113
Gene Name
Zic family member 4
Gene Aliases
-
Chromosome Location
Chr.1: 262545401 - 262563114 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959518.1 XP_027815319.1 3 - 3 410 94

View Full Product Details