Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC101115495
Entrez Gene ID
101115495
Gene Name
C-reactive protein
Gene Aliases
-
Chromosome Location
Chr.1: 117562417 - 117565817 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002651.4 XP_004002700.1 1 - 2 175 96

View Full Product Details