Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INO80C
Entrez Gene ID
101110106
Gene Name
INO80 complex subunit C
Gene Aliases
-
Chromosome Location
Chr.23: 25874426 - 25897590 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015103598.2 XP_014959084.1 4 - 5 395 95
XM_027960718.1 XP_027816519.1 4 - 5 618 95
XM_027960719.1 XP_027816520.1 5 - 6 741 95

View Full Product Details