Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SALL4
Entrez Gene ID
101119746
Gene Name
spalt like transcription factor 4
Gene Aliases
-
Chromosome Location
Chr.13: 82988671 - 83005772 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977337.1 XP_027833138.1 2 - 3 2491 86

View Full Product Details