Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AQP10
Entrez Gene ID
101102866
Gene Name
aquaporin 10
Gene Aliases
-
Chromosome Location
Chr.1: 110732977 - 110736483 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003657.4 XP_004003706.1 5 - 6 743 73

View Full Product Details