Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUNDC1
Entrez Gene ID
101107804
Gene Name
RUN domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 20088992 - 20097681 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012954.4 XP_004013003.1 1 - 2 500 95

View Full Product Details