Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLYATL2
Entrez Gene ID
101109387
Gene Name
glycine-N-acyltransferase like 2
Gene Aliases
-
Chromosome Location
Chr.15: 88556182 - 88567869 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016498.4 XP_004016547.1 2 - 3 346 82

View Full Product Details