Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC10B
Entrez Gene ID
101109402
Gene Name
leucine rich repeat containing 10B
Gene Aliases
-
Chromosome Location
Chr.21: 41861164 - 41863149 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959351.1 XP_027815152.1 1 - 1 767 90

View Full Product Details