Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D7
Entrez Gene ID
101114647
Gene Name
SH2 domain containing 7
Gene Aliases
-
Chromosome Location
Chr.18: 28420732 - 28431251 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018087.4 XP_004018136.1 2 - 3 274 69

View Full Product Details