Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AARD
Entrez Gene ID
101120843
Gene Name
alanine and arginine rich domain containing protein
Gene Aliases
-
Chromosome Location
Chr.9: 66765430 - 66769944 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011989.4 XP_004012038.1 1 - 2 316 85

View Full Product Details