Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC105602020
Entrez Gene ID
105602020
Gene Name
zinc finger protein 28 homolog
Gene Aliases
ZFP28
Chromosome Location
Chr.14: 68862968 - 68945746 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978006.1 XP_027833807.1 6 - 7 1050 109

View Full Product Details