Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THOC3
Entrez Gene ID
101103479
Gene Name
THO complex 3
Gene Aliases
-
Chromosome Location
Chr.7: 5960156 - 5972493 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096786.2 XP_014952272.2 1 - 2 366 115

View Full Product Details