Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUPT7L
Entrez Gene ID
101120226
Gene Name
SPT7 like, STAGA complex subunit gamma
Gene Aliases
-
Chromosome Location
Chr.3: 36207156 - 36221428 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005776.4 XP_004005825.1 4 - 5 1429 94
XM_012166878.3 XP_012022268.1 5 - 6 1507 94

View Full Product Details