Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT3A
Entrez Gene ID
101123374
Gene Name
Wnt family member 3A
Gene Aliases
-
Chromosome Location
Chr.5: 3037287 - 3111535 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969327.1 XP_027825128.1 3 - 4 582 75

View Full Product Details